Gene Transcripts

Gene Symbol
ZFAND1
Entrez Gene ID
79752
Gene Name
zinc finger AN1-type containing 1
Gene Aliases
Chromosome Location
Chr. 8: 81701331 - 81721304 on Build GRCh38
UniGene ID
Hs.655453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170796.1 NP_001164267.1 2, 3 119
NM_001170797.1 NP_001164268.1 2, 3 119
NM_024699.2 NP_078975.2 2, 3 119
GenBank mRNA AK024069.1 - 2, 3 110
AK225195.1 - 2, 3 115
BC022251.2 - 2, 3 104 4695118
BQ427843.1 - 2, 3 119
CR457323.1 - 2, 3 96

View Full Product Details