Gene Transcripts

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location
Chr. 10: 32446082 - 32882864 on Build GRCh38
UniGene ID
Hs.585464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321115.1 NP_001308044.1 24 2919
XM_005252593.2 XP_005252650.1 5 706
XM_006717505.1 XP_006717568.1 22 2414
XM_006717506.1 XP_006717569.1 21 2306
XM_011519663.1 XP_011517965.1 24 2627
XM_011519664.1 XP_011517966.1 24 2627
XM_011519666.1 XP_011517968.1 24 2627
XM_011519667.1 XP_011517969.1 24 2627
XM_011519668.1 XP_011517970.1 23 2537
XM_011519669.1 XP_011517971.1 23 2525
XM_011519670.1 XP_011517972.1 23 2519
XM_011519671.1 XP_011517973.1 24 2627
XM_011519673.1 XP_011517975.1 24 2627
XM_011519674.1 XP_011517976.1 24 2627
XM_011519675.1 XP_011517977.1 22 2417
XM_011519676.1 XP_011517978.1 24 2627
XM_011519677.1 XP_011517979.1 23 2405
XM_011519679.1 XP_011517981.1 21 2096
XM_011519680.2 XP_011517982.1 19 1762
XM_011519683.2 XP_011517985.1 13 1436
XM_011519684.2 XP_011517986.1 24 2627
XM_011519685.1 XP_011517987.1 16 1644
XM_011519686.2 XP_011517988.1 24 2627
XM_011519687.1 XP_011517989.1 6 551
XM_017016638.1 XP_016872127.1 24 2627
XM_017016639.1 XP_016872128.1 24 2627
XM_017016640.1 XP_016872129.1 24 2627
XM_017016641.1 XP_016872130.1 19 2056
XM_017016642.1 XP_016872131.1 19 2129
XM_017016643.1 XP_016872132.1 19 1746
XM_017016644.1 XP_016872133.1 24 2627
XM_017016645.1 XP_016872134.1 18 1670
XM_017016646.1 XP_016872135.1 15 1557
XM_017016647.1 XP_016872136.1 24 2627
XM_017016648.1 XP_016872137.1 11 1278
XM_017016649.1 XP_016872138.1 11 1218
XM_017016650.1 XP_016872139.1 24 2627
XM_017016651.1 XP_016872140.1 24 2627
XM_017016652.1 XP_016872141.1 8 681
XM_017016653.1 XP_016872142.1 6 591
GenBank mRNA AK023093.1 - 5 712
AK301824.1 - 2 147
BC125090.1 - 8 881 40118176
BC125091.1 - 4 351 40118177
DB026046.1 - 1 65

View Full Product Details