Gene Transcripts

Gene Symbol
ZBTB16
Entrez Gene ID
7704
Gene Name
zinc finger and BTB domain containing 16
Gene Aliases
PLZF, ZNF145
Chromosome Location
Chr. 11: 114059576 - 114256765 on Build GRCh38
UniGene ID
Hs.591945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018011.1 NP_001018011.1 2 462
NM_006006.4 NP_005997.2 2 617
XM_005271658.4 XP_005271715.1 2 749
XM_017018257.1 XP_016873746.1 2 550
XM_017018258.1 XP_016873747.1 2 705
XM_017018259.1 XP_016873748.1 2 1057
GenBank mRNA AB208916.1 - 2 601
BC026902.1 - 2 521 4944546
BC029812.1 - 2 599 5175961
BX648973.1 - 2 734
KU178400.1 - 1 353
KU178401.1 - 1 353
Z19002.1 - 1 428

View Full Product Details