Gene Transcripts

Gene Symbol
PYROXD1
Entrez Gene ID
79912
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Gene Aliases
Chromosome Location
Chr. 12: 21437604 - 21471250 on Build GRCh38
UniGene ID
Hs.709545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024854.3 NP_079130.2 4, 5 549
XM_006719153.3 XP_006719216.1 4, 5 523
GenBank mRNA AK025681.1 - 4, 5 451
AK125461.1 - 4, 5 962
AL832441.1 - 4, 5 476
BC021662.1 - 4, 5 450 3826938
DB041552.1 - 4, 5 549

View Full Product Details