Gene Transcripts

Gene Symbol
RPP21
Entrez Gene ID
79897
Gene Name
ribonuclease P/MRP subunit p21
Gene Aliases
C6orf135, CAT60
Chromosome Location
Chr. 6: 30345129 - 30346858 on Build GRCh38
UniGene ID
Hs.183232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199120.1 NP_001186049.1 5 522
NM_001199121.1 NP_001186050.1 5 505
NM_024839.2 NP_079115.1 5 498
GenBank mRNA AF212152.2 - 5 468
AJ504713.1 - 5 458
AJ504714.1 - 4 537
AJ504715.1 - 4 527
AJ504716.1 - 5 465
AK026291.1 - 5 467
BC011730.2 - 5 460 4422871
BG708140.1 - 5 529
BI598757.1 - 5 525
DA270919.1 - 5 498

Gene Symbol
TRIM39-RPP21
Entrez Gene ID
202658
Gene Name
TRIM39-RPP21 readthrough
Gene Aliases
TRIM39R
Chromosome Location
Chr. 6: 30329311 - 30346858 on Build GRCh38
UniGene ID
Hs.413493
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199119.1 NP_001186048.1 10 1509
GenBank mRNA EU012025.1 - 10 1238

View Full Product Details