Gene Transcripts

Gene Symbol
NYNRIN
Entrez Gene ID
57523
Gene Name
NYN domain and retroviral integrase containing
Gene Aliases
CGIN1, KIAA1305
Chromosome Location
Chr. 14: 24398786 - 24419288 on Build GRCh38
UniGene ID
Hs.288348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025081.2 NP_079357.2 9 4880
XM_011537016.2 XP_011535318.1 4 3155
GenBank mRNA AB037726.2 - 7 4325
AK021873.1 - 3 665

View Full Product Details