Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308445.1 | NP_001295374.1 | 2 | 427 | |
NM_025146.3 | NP_079422.1 | 2 | 430 | ||
GenBank mRNA | AK023090.1 | - | 2 | 345 | |
AK023256.1 | - | 2 | 380 | ||
AK296456.1 | - | 2 | 125 | ||
AK315876.1 | - | 2 | 232 | ||
AL601126.1 | - | 2 | 325 | ||
BC012731.2 | - | 2 | 369 | 3619903 | |
BM831426.1 | - | 2 | 404 | ||
CR749314.1 | - | 2 | 325 | ||
DA854531.1 | - | 2 | 427 |