Gene Transcripts

Gene Symbol
COQ10B
Entrez Gene ID
80219
Gene Name
coenzyme Q10B
Gene Aliases
Chromosome Location
Chr. 2: 197453423 - 197475309 on Build GRCh38
UniGene ID
Hs.606592
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320818.1 NP_001307747.1 2 263
NM_001320819.1 NP_001307748.1 3 457
NM_001320820.1 NP_001307749.1 3 654
NM_025147.4 NP_079423.1 3 413
GenBank mRNA AK023510.1 - 3 282
AK294093.1 - 3 654
AK308977.1 - 3 251
BC022453.1 - 3 330 4796819
BU195968.1 - 2 197
DC328985.1 - 3 251
DC349756.1 - 3 414
HY088880.1 - 3 406

View Full Product Details