Gene Transcripts

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
Chromosome Location
Chr. 1: 1292384 - 1307925 on Build GRCh38
UniGene ID
Hs.535257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030649.2 NP_085152.2 4 343
XM_005244715.2 XP_005244772.1 4 379
XM_011540606.2 XP_011538908.1 4 392
XM_011540607.1 XP_011538909.1 4 2816
XM_011540608.2 XP_011538910.1 4 393
XM_011540609.2 XP_011538911.1 4 393
XM_017000233.1 XP_016855722.1 4 294
GenBank mRNA AB051503.1 - 3 2008
AF411981.1 - 2 143
AK093954.1 - 4 390
AK098490.1 - 3 2547
BC051194.1 - 4 343 4819254

View Full Product Details