Gene Transcripts

Gene Symbol
PITPNM3
Entrez Gene ID
83394
Gene Name
PITPNM family member 3
Gene Aliases
ACKR6, CORD5, NIR1, RDGBA3
Chromosome Location
Chr. 17: 6451263 - 6556557 on Build GRCh38
UniGene ID
Hs.183983
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031220.3 NP_112497.2 3 292
XM_011524015.2 XP_011522317.1 3 232
XM_011524016.2 XP_011522318.1 3 233
XM_011524017.2 XP_011522319.1 3 234
GenBank mRNA AF334586.1 - 3 289
AL389994.1 - 1 15
BC128583.1 - 3 229 40128716
BC128584.1 - 3 229 40128724

View Full Product Details