Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005334.2 | NP_005325.2 | 22 | 5960 | |
XM_006724815.2 | XP_006724878.1 | 22 | 6560 | ||
XM_006724816.2 | XP_006724879.1 | 22 | 6557 | ||
XM_011531147.2 | XP_011529449.1 | 22 | 6437 | ||
XM_011531148.2 | XP_011529450.1 | 22 | 6434 | ||
XM_017029471.1 | XP_016884960.1 | 21 | 6359 | ||
XM_017029472.1 | XP_016884961.1 | 17 | 4745 | ||
GenBank mRNA | L20010.1 | - | 22 | 5776 |