Gene Transcripts
- Gene Symbol
- GORASP1
- Entrez Gene ID
- 64689
- Gene Name
- golgi reassembly stacking protein 1
- Gene Aliases
- GOLPH5, GRASP65, P65
- Chromosome Location
- Chr. 3: 39096599 - 39108363 on Build GRCh38
- UniGene ID
- Hs.721946
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031899.3 |
NP_114105.1
|
2, 3 |
970 |
|
NR_103866.1 |
|
2, 3 |
507 |
|
NR_103867.1 |
|
2, 3 |
507 |
|
XM_006713301.3 |
XP_006713364.1
|
2, 3 |
1070 |
|
XM_011534020.2 |
XP_011532322.1
|
2, 3 |
1070 |
|
XM_011534021.2 |
XP_011532323.1
|
2, 3 |
1070 |
|
XM_017007050.1 |
XP_016862539.1
|
2, 3 |
246 |
|
XM_017007051.1 |
XP_016862540.1
|
2, 3 |
685 |
|
XM_017007052.1 |
XP_016862541.1
|
2, 3 |
232 |
|
GenBank mRNA |
AJ409349.1 |
- |
2, 3 |
246 |
|
AK001574.1 |
- |
2, 3 |
245 |
|
AK091168.1 |
- |
2, 3 |
434 |
|
AK124755.1 |
- |
2, 3 |
970 |
|
AK300271.1 |
- |
2, 3 |
186 |
|
BC008928.2 |
- |
2, 3 |
228 |
|
BC075854.1 |
- |
2, 3 |
256 |
5503263 |
BC103826.1 |
- |
2, 3 |
188 |
40001584 |
BC103827.1 |
- |
2, 3 |
188 |
40001588 |
DB031221.1 |
- |
2, 3 |
245 |
|
View Full Product Details