Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031308.3 | NP_112598.3 | 2 | 2031 | |
XM_017013889.1 | XP_016869378.1 | 2 | 2063 | ||
XM_017013890.1 | XP_016869379.1 | 2 | 2202 | ||
XM_017013891.1 | XP_016869380.1 | 2 | 2063 | ||
XM_017013892.1 | XP_016869381.1 | 2 | 2063 | ||
GenBank mRNA | AB051895.1 | - | 1 | 1972 |