Gene Transcripts

Gene Symbol
TTC25
Entrez Gene ID
83538
Gene Name
tetratricopeptide repeat domain 25
Gene Aliases
Chromosome Location
Chr. 17: 41930605 - 41966517 on Build GRCh38
UniGene ID
Hs.201134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031421.3 NP_113609.1 12 2116
NR_110662.1 10 1683
XM_011525323.2 XP_011523625.1 12 2056
XM_011525324.2 XP_011523626.1 11 1982
GenBank mRNA AK055498.1 - 12 2061
AL136760.1 - 12 2083
BC025390.1 - 10 1691
BC067297.1 - 12 2076 30383608

View Full Product Details