Gene Transcripts

Gene Symbol
C3orf20
Entrez Gene ID
84077
Gene Name
chromosome 3 open reading frame 20
Gene Aliases
Chromosome Location
Chr. 3: 14673532 - 14773036 on Build GRCh38
UniGene ID
Hs.661452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184957.1 NP_001171886.1 3 420
NM_001184958.1 NP_001171887.1 3 525
NM_032137.4 NP_115513.4 3 922
XM_006713341.2 XP_006713404.1 3 922
XM_006713342.2 XP_006713405.1 3 775
XM_006713343.2 XP_006713406.1 3 922
XM_011534153.2 XP_011532455.1 3 2552
XM_011534154.2 XP_011532456.1 3 1987
XM_011534158.2 XP_011532460.1 3 922
XM_017007310.1 XP_016862799.1 3 922
GenBank mRNA AK131481.1 - 3 776
AL136781.1 - 3 340
AL834386.1 - 3 874
BC030599.2 - 3 483 4831210
BC038406.1 - 3 842 5167835
BG718869.1 - 3 488

View Full Product Details