Gene Transcripts

Gene Symbol
CDCA7
Entrez Gene ID
83879
Gene Name
cell division cycle associated 7
Gene Aliases
ICF3, JPO1
Chromosome Location
Chr. 2: 173354017 - 173368992 on Build GRCh38
UniGene ID
Hs.470654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031942.4 NP_114148.3 6 973
NM_145810.2 NP_665809.1 5 736
XM_011511957.2 XP_011510259.1 6 1514
GenBank mRNA AK027628.1 - 6 945
AK027642.1 - 5 717
AK075134.1 - 4 1599
AK223524.1 - 5 741
AK297097.1 - 4 571
AK297245.1 - 6 833
AK300949.1 - 6 802
AL833728.1 - 1 265
AL834186.1 - 4 1578
AY029179.1 - 5 749
BC015124.1 - 1 152
BC027966.1 - 5 702 5227112

View Full Product Details