Gene Transcripts

Gene Symbol
FGFBP2
Entrez Gene ID
83888
Gene Name
fibroblast growth factor binding protein 2
Gene Aliases
HBP17RP, KSP37
Chromosome Location
Chr. 4: 15960240 - 15963236 on Build GRCh38
UniGene ID
Hs.98785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031950.3 NP_114156.1 1 230
GenBank mRNA AB021123.1 - 1 201
AF361936.1 - 1 123
AY358978.1 - 1 230
BC025720.1 - 1 199 5228917

View Full Product Details