Gene Transcripts
- Gene Symbol
- TEX35
- Entrez Gene ID
- 84066
- Gene Name
- testis expressed 35
- Gene Aliases
- C1orf49, TSC24
- Chromosome Location
- Chr. 1: 178513077 - 178523500 on Build GRCh38
- UniGene ID
- Hs.534501
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170722.1 |
NP_001164193.1
|
4, 5 |
348 |
|
NM_001170723.1 |
NP_001164194.1
|
4, 5 |
324 |
|
NM_001170724.1 |
NP_001164195.1
|
4, 5 |
324 |
|
NM_032126.4 |
NP_115502.2
|
4, 5 |
324 |
|
GenBank mRNA |
AK093269.1 |
- |
4, 5 |
271 |
|
AL136694.1 |
- |
4, 5 |
328 |
|
AL552172.3 |
- |
4, 5 |
246 |
|
BC034972.2 |
- |
4, 5 |
281 |
4823687 |
BI461686.1 |
- |
4, 5 |
289 |
|
BI825610.1 |
- |
4, 5 |
258 |
|
View Full Product Details