Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032167.4 | NP_115543.3 | 3 | 177 | |
XM_005255682.3 | XP_005255739.1 | 3 | 202 | ||
XM_005255683.3 | XP_005255740.1 | 3 | 218 | ||
XM_011522738.2 | XP_011521040.1 | 3 | 349 | ||
XM_011522741.2 | XP_011521043.1 | 3 | 333 | ||
XM_011522743.2 | XP_011521045.1 | 3 | 337 | ||
XM_011522744.2 | XP_011521046.1 | 3 | 208 | ||
XM_017023871.1 | XP_016879360.1 | 3 | 346 | ||
XM_017023872.1 | XP_016879361.1 | 3 | 166 | ||
XM_017023873.1 | XP_016879362.1 | 3 | 333 | ||
XM_017023874.1 | XP_016879363.1 | 3 | 166 | ||
XM_017023875.1 | XP_016879364.1 | 2 | 54 | ||
XM_017023876.1 | XP_016879365.1 | 3 | 211 | ||
XM_017023877.1 | XP_016879366.1 | 3 | 166 | ||
GenBank mRNA | AK022425.1 | - | 3 | 166 | |
AL135642.1 | - | 3 | 166 | ||
HY078299.1 | - | 3 | 178 |