Gene Transcripts

Gene Symbol
C9orf64
Entrez Gene ID
84267
Gene Name
chromosome 9 open reading frame 64
Gene Aliases
Chromosome Location
Chr. 9: 83938311 - 83956986 on Build GRCh38
UniGene ID
Hs.208914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317997.1 NP_001304926.1 4 516
NM_032307.4 NP_115683.3 3 914
GenBank mRNA AK090882.1 - 4 516
BC004407.1 - 3 889
BC137461.1 - 3 791 9021461
BC137462.1 - 3 791 9021462

View Full Product Details