Gene Transcripts

Gene Symbol
CCDC77
Entrez Gene ID
84318
Gene Name
coiled-coil domain containing 77
Gene Aliases
Chromosome Location
Chr. 12: 389350 - 442642 on Build GRCh38
UniGene ID
Hs.631656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130146.1 NP_001123618.1 3 302
NM_001130147.1 NP_001123619.1 3 329
NM_001130148.1 NP_001123620.1 2 275
NM_032358.3 NP_115734.1 4 356
GenBank mRNA AK027638.1 - 4 274
AK293165.1 - 3 240
AK308216.1 - 3 284
AK316029.1 - 2 277
BC006444.2 - 3 245 3616619
CX784327.1 - 3 209
DA711064.1 - 4 357
DC331633.1 - 2 275

View Full Product Details