Gene Transcripts

Gene Symbol
CARD19
Entrez Gene ID
84270
Gene Name
caspase recruitment domain family member 19
Gene Aliases
BinCARD, C9orf89
Chromosome Location
Chr. 9: 93096123 - 93113295 on Build GRCh38
UniGene ID
Hs.434213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318010.1 NP_001304939.1 4 723
NM_001318011.1 NP_001304940.1 5 866
NM_032310.4 NP_115686.3 6 926
NR_134464.1 4 2373
NR_134465.1 4 2362
NR_134466.1 6 937
NR_134467.1 7 1028
NR_134468.1 5 877
GenBank mRNA AK057716.1 - 4 2267
AK091611.1 - 4 2278
AK097977.1 - 2 2664
AK128259.1 - 2 3399
BC004500.2 - 7 795 3833318
BC038856.1 - 4 2254 5215640
BC091476.1 - 6 780
DB550397.1 - 4 422
DN998463.1 - 4 616

View Full Product Details