Gene Transcripts
- Gene Symbol
- MFSD14B
- Entrez Gene ID
- 84641
- Gene Name
- major facilitator superfamily domain containing 14B
- Gene Aliases
- HIATL1
- Chromosome Location
- Chr. 9: 94374327 - 94460920 on Build GRCh38
- UniGene ID
- Hs.555996
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032558.2 |
NP_115947.2
|
3, 4 |
544 |
|
XM_006717305.3 |
XP_006717368.1
|
3, 4 |
519 |
|
XM_011519114.2 |
XP_011517416.1
|
3, 4 |
235 |
|
XM_017015217.1 |
XP_016870706.1
|
3, 4 |
345 |
|
XM_017015218.1 |
XP_016870707.1
|
3, 4 |
272 |
|
XM_017015219.1 |
XP_016870708.1
|
3, 4 |
301 |
|
XM_017015220.1 |
XP_016870709.1
|
3, 4 |
211 |
|
XM_017015221.1 |
XP_016870710.1
|
3, 4 |
235 |
|
GenBank mRNA |
AK300615.1 |
- |
2, 3 |
116 |
|
BC106062.1 |
- |
3, 4 |
281 |
6162855 |
BU507687.1 |
- |
3, 4 |
521 |
|
BX359483.2 |
- |
3, 4 |
551 |
|
- Gene Symbol
- MFSD14C
- Entrez Gene ID
- 84278
- Gene Name
- major facilitator superfamily domain containing 14C
- Gene Aliases
- HIATL2
- Chromosome Location
- Chr. 9: 96946045 - 97013580 on Build GRCh38
- UniGene ID
- Hs.610084
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002894.1 |
|
3, 4 |
489 |
|
GenBank mRNA |
BC005058.1 |
- |
3, 4 |
489 |
|
View Full Product Details