Gene Transcripts

Gene Symbol
ACBD6
Entrez Gene ID
84320
Gene Name
acyl-CoA binding domain containing 6
Gene Aliases
Chromosome Location
Chr. 1: 180269663 - 180502967 on Build GRCh38
UniGene ID
Hs.200051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032360.3 NP_115736.1 4 1029
GenBank mRNA AK314112.1 - 4 1004
AY606133.1 - 4 443
BC006505.2 - 4 486 2821736

View Full Product Details