Gene Transcripts
- Gene Symbol
- ACSS1
- Entrez Gene ID
- 84532
- Gene Name
- acyl-CoA synthetase short-chain family member 1
- Gene Aliases
- ACAS2L, ACECS1, AceCS2L
- Chromosome Location
- Chr. 20: 25006230 - 25058182 on Build GRCh38
- UniGene ID
- Hs.529353
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252675.1 |
NP_001239604.1
|
2, 3 |
507 |
|
NM_001252677.1 |
NP_001239606.1
|
2, 3 |
507 |
|
NM_032501.3 |
NP_115890.2
|
2, 3 |
507 |
|
XM_006723659.1 |
XP_006723722.1
|
2, 3 |
468 |
|
XM_011529388.1 |
XP_011527690.1
|
2, 3 |
468 |
|
GenBank mRNA |
AK127566.1 |
- |
2, 3 |
453 |
|
AK314440.1 |
- |
2, 3 |
430 |
|
AL832939.1 |
- |
2, 3 |
191 |
|
BC039261.1 |
- |
2, 3 |
466 |
5298849 |
BC044588.1 |
- |
2, 3 |
432 |
5260496 |
BC055008.1 |
- |
2, 3 |
432 |
4838976 |
BC114612.1 |
- |
2, 3 |
298 |
|
BM150326.1 |
- |
2, 3 |
507 |
|
View Full Product Details