Gene Transcripts

Gene Symbol
ACSS1
Entrez Gene ID
84532
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
ACAS2L, ACECS1, AceCS2L
Chromosome Location
Chr. 20: 25006230 - 25058182 on Build GRCh38
UniGene ID
Hs.529353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252675.1 NP_001239604.1 7 1221
NM_001252676.1 NP_001239605.1 6 1048
NM_001252677.1 NP_001239606.1 7 1221
NM_032501.3 NP_115890.2 7 1221
XM_011529388.1 XP_011527690.1 6 1006
GenBank mRNA AB058749.1 - 2 138
AK024396.1 - 1 2255
AK024424.1 - 1 1782
AK027817.1 - 1 44
AK092295.1 - 5 630
AK125058.1 - 5 2424
AK127566.1 - 7 1167
AK296306.1 - 6 1045
AK314440.1 - 7 1144
BC039261.1 - 7 1180 5298849
BC044588.1 - 7 1146 5260496
BC055008.1 - 7 1146 4838976
BC114612.1 - 7 1012

View Full Product Details