Gene Transcripts

Gene Symbol
CRACR2A
Entrez Gene ID
84766
Gene Name
calcium release activated channel regulator 2A
Gene Aliases
EFCAB4B
Chromosome Location
Chr. 12: 3613366 - 3753216 on Build GRCh38
UniGene ID
Hs.504534
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144958.1 NP_001138430.1 4 697
NM_032680.3 NP_116069.1 4 697
XM_006719021.3 XP_006719084.1 4 697
XM_011521034.2 XP_011519336.1 3 444
XM_011521036.2 XP_011519338.1 2 204
GenBank mRNA AK304017.1 - 4 599
BC004524.2 - 4 594
BC150643.1 - 3 328 9057013

View Full Product Details