Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017829.5 | NP_060299.4 | 8 | 1556 | |
NM_033070.2 | NP_149061.1 | 8 | 1631 | ||
XM_005261236.3 | XP_005261293.1 | 8 | 1609 | ||
XM_011546126.2 | XP_011544428.1 | 7 | 1427 | ||
XM_011546127.1 | XP_011544429.1 | 8 | 2046 | ||
XM_017028781.1 | XP_016884270.1 | 8 | 2098 | ||
XM_017028782.1 | XP_016884271.1 | 9 | 2425 | ||
XM_017028783.1 | XP_016884272.1 | 9 | 2862 | ||
XM_017028784.1 | XP_016884273.1 | 9 | 2425 | ||
GenBank mRNA | AF273270.1 | - | 8 | 1556 | |
AF273271.1 | - | 8 | 1616 | ||
AK000461.1 | - | 8 | 1629 | ||
AK001034.1 | - | 8 | 1608 | ||
AK123646.1 | - | 3 | 2112 | ||
AL833450.1 | - | 3 | 3447 | ||
BC042540.1 | - | 8 | 1615 | 4342582 |