Gene Transcripts

Gene Symbol
HSPA12B
Entrez Gene ID
116835
Gene Name
heat shock protein family A (Hsp70) member 12B
Gene Aliases
C20orf60
Chromosome Location
Chr. 20: 3732670 - 3753111 on Build GRCh38
UniGene ID
Hs.516854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197327.1 NP_001184256.1 10 1170
NM_001318322.1 NP_001305251.1 9 1075
NM_052970.4 NP_443202.3 10 1173
XM_011529151.2 XP_011527453.1 7 871
XM_017027632.1 XP_016883121.1 10 1088
XM_017027633.1 XP_016883122.1 10 1072
XM_017027634.1 XP_016883123.1 10 1072
XM_017027635.1 XP_016883124.1 10 1116
GenBank mRNA AK056712.1 - 10 1110
AK297120.1 - 8 966
AK301294.1 - 9 1041
BC090857.1 - 9 1034
BC110881.1 - 10 1122 5725594
BC117284.1 - 10 1071 40125835
BC126263.1 - 10 1071 8991979
BC143932.1 - 10 1068 9052450

View Full Product Details