Gene Transcripts

Gene Symbol
EVA1C
Entrez Gene ID
59271
Gene Name
eva-1 homolog C (C. elegans)
Gene Aliases
B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21
Chromosome Location
Chr. 21: 32411692 - 32515400 on Build GRCh38
UniGene ID
Hs.208358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286556.1 NP_001273485.1 3 864
NM_001320745.1 NP_001307674.1 3 789
NM_058187.4 NP_478067.2 3 864
NR_104472.1 2 667
NR_135467.1 3 920
XM_005261019.4 XP_005261076.1 3 985
XM_006724038.3 XP_006724101.1 3 985
XM_011529669.2 XP_011527971.1 3 1116
XM_017028418.1 XP_016883907.1 3 982
XM_017028419.1 XP_016883908.1 3 985
XM_017028420.1 XP_016883909.1 4 905
XM_017028421.1 XP_016883910.1 3 789
GenBank mRNA AF358258.1 - 3 447
AK125013.1 - 2 1414
AK126660.1 - 1 3988
AL543462.3 - 2 666
AY040087.1 - 2 250
AY358787.1 - 3 857
BC038710.1 - 3 599 4813873
BM836911.1 - 2 192

View Full Product Details