Gene Transcripts

Gene Symbol
CELF3
Entrez Gene ID
11189
Gene Name
CUGBP, Elav-like family member 3
Gene Aliases
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4
Chromosome Location
Chr. 1: 151700058 - 151716821 on Build GRCh38
UniGene ID
Hs.26047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172648.3 NP_001166119.1 13 2447
NM_001172649.3 NP_001166120.1 12 2544
NM_001291106.1 NP_001278035.1 13 2691
NM_001291107.1 NP_001278036.1 13 2691
NM_007185.6 NP_009116.3 13 2694
XM_006711139.2 XP_006711202.1 14 1922
XM_006711141.2 XP_006711204.1 14 1919
XM_011509111.2 XP_011507413.1 12 2389
XM_011509112.2 XP_011507414.1 12 1785
XM_011509114.2 XP_011507416.1 12 1753
XM_017000145.1 XP_016855634.1 12 1760
XM_017000146.1 XP_016855635.1 12 1757
GenBank mRNA AB209264.1 - 13 2447
AF284423.1 - 4 921
AK298852.1 - 7 1317
AY165003.1 - 13 2684
BC004145.2 - 9 1070
BC041088.1 - 12 1776
BC052491.1 - 13 2665 4820737
BC068008.1 - 12 1721
U80746.1 - 10 1573
U80759.1 - 10 1580

View Full Product Details