Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_130446.2 | NP_569713.2 | 1 | 172 | |
XM_011513274.2 | XP_011511576.1 | 1 | 173 | ||
GenBank mRNA | AF441792.1 | - | 1 | 84 | |
AK097125.1 | - | 1 | 126 | ||
AK314473.1 | - | 1 | 126 | ||
BC032348.1 | - | 1 | 150 | 5213743 | |
BP284594.1 | - | 1 | 172 | ||
BX648370.1 | - | 1 | 127 |