Gene Transcripts
- Gene Symbol
- CPNE4
- Entrez Gene ID
- 131034
- Gene Name
- copine 4
- Gene Aliases
- COPN4, CPN4
- Chromosome Location
- Chr. 3: 131533560 - 132285702 on Build GRCh38
- UniGene ID
- Hs.199877
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289112.1 |
NP_001276041.1
|
6, 7 |
848 |
|
NM_130808.2 |
NP_570720.1
|
6, 7 |
1476 |
|
NM_153429.1 |
NP_702907.1
|
7, 8 |
836 |
|
XM_011512406.2 |
XP_011510708.1
|
5, 6 |
623 |
|
XM_011512407.2 |
XP_011510709.1
|
5, 6 |
880 |
|
XM_011512408.2 |
XP_011510710.1
|
5, 6 |
2008 |
|
XM_017005693.1 |
XP_016861182.1
|
9, 10 |
2229 |
|
XM_017005694.1 |
XP_016861183.1
|
5, 6 |
600 |
|
XM_017005695.1 |
XP_016861184.1
|
6, 7 |
869 |
|
GenBank mRNA |
AF465770.1 |
- |
9, 10 |
1176 |
|
AF465771.1 |
- |
7, 8 |
836 |
|
AK056175.1 |
- |
6, 7 |
948 |
|
AK128117.1 |
- |
10, 11 |
2726 |
|
AK295557.1 |
- |
6, 7 |
763 |
|
BC014396.1 |
- |
6, 7 |
1034 |
3162811 |
BC075046.2 |
- |
6, 7 |
625 |
30915576 |
View Full Product Details