Gene Transcripts
- Gene Symbol
- XAGE1B
- Entrez Gene ID
- 653220
- Gene Name
- X antigen family member 1B
- Gene Aliases
- CT12.1, CT12.1A, CT12.1B, CTP9, GAGED2, XAGE-1, XAGE1, XAGE1A
- Chromosome Location
- Chr. X: 52495668 - 52500815 on Build GRCh38
- UniGene ID
- Hs.112208
- Species
- Homo sapiens
- Gene Symbol
- XAGE1E
- Entrez Gene ID
- 653067
- Gene Name
- X antigen family member 1E
- Gene Aliases
- CT12.1, CT12.1C, CT12.1D, CT12.1E, CTP9, GAGED2, XAGE-1, XAGE1, XAGE1C, XAGE1D
- Chromosome Location
- Chr. X: 52512074 - 52517221 on Build GRCh38
- UniGene ID
- Hs.112208
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001097604.2 |
NP_001091073.2
|
3, 4 |
519 |
|
NM_001097605.2 |
NP_001091074.1
|
3, 4 |
382 |
|
NR_033257.1 |
|
3, 4 |
630 |
|
XM_017029746.1 |
XP_016885235.1
|
3, 4 |
651 |
|
XM_017029747.1 |
XP_016885236.1
|
3, 4 |
519 |
|
GenBank mRNA |
AF251237.1 |
- |
3, 4 |
511 |
|
AJ290447.1 |
- |
3, 4 |
355 |
|
AJ318878.1 |
- |
3, 4 |
630 |
|
AJ318879.1 |
- |
3, 4 |
371 |
|
BC009538.1 |
- |
3, 4 |
414 |
|
BT007099.1 |
- |
2, 3 |
231 |
|
View Full Product Details