Gene Transcripts

Gene Symbol
HNRNPLL
Entrez Gene ID
92906
Gene Name
heterogeneous nuclear ribonucleoprotein L like
Gene Aliases
HNRPLL, SRRF
Chromosome Location
Chr. 2: 38563186 - 38603036 on Build GRCh38
UniGene ID
Hs.445497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142650.1 NP_001136122.1 2, 3 354
NM_138394.3 NP_612403.2 1, 2 610
XM_005264639.3 XP_005264696.2 1, 2 607
XM_005264640.3 XP_005264697.2 2, 3 353
XM_011533165.2 XP_011531467.1 1, 2 607
XM_011533166.2 XP_011531468.1 1, 2 607
GenBank mRNA AK000155.1 - 2, 3 258
AK291462.1 - 1, 2 475
AK292259.1 - 2, 3 347
AY236962.1 - 1, 2 439
AY236963.2 - 2, 3 199
BC008217.1 - 2, 3 247 3449607
BC017480.1 - 1, 2 456 3502860
DA760455.1 - 3, 4 290
KU178839.1 - 1, 2 200
KU178840.1 - 1, 2 200
KU178841.1 - 1, 2 200

View Full Product Details