Gene Transcripts

Gene Symbol
MTFR2
Entrez Gene ID
113115
Gene Name
mitochondrial fission regulator 2
Gene Aliases
DUFD1, FAM54A
Chromosome Location
Chr. 6: 136231030 - 136253659 on Build GRCh38
UniGene ID
Hs.121536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099286.2 NP_001092756.1 6 1029
NM_001318738.1 NP_001305667.1 7 841
NM_138419.4 NP_612428.2 6 741
XM_011535410.2 XP_011533712.1 7 1093
XM_011535412.2 XP_011533714.1 5 834
XM_011535413.2 XP_011533715.1 5 3948
GenBank mRNA AK125758.1 - 7 798
AK292303.1 - 6 980
BC011716.2 - 3 364
BC063688.1 - 6 694 4455541

View Full Product Details