Gene Transcripts

Gene Symbol
TM4SF19
Entrez Gene ID
116211
Gene Name
transmembrane 4 L six family member 19
Gene Aliases
OCTM4
Chromosome Location
Chr. 3: 196323546 - 196338420 on Build GRCh38
UniGene ID
Hs.733264
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204897.1 NP_001191826.1 5 795
NM_001204898.1 NP_001191827.1 4 721
NM_138461.3 NP_612470.2 5 799
GenBank mRNA AF192910.1 - 5 755
AK310209.1 - 5 765
BC146985.1 - 5 700 9056913
BC146993.1 - 5 700 9056921
BC147021.1 - 5 700 9056949
BC171870.1 - 4 622 9054524
BP395549.1 - 2 239
BP396791.1 - 2 240
CA489370.1 - 4 721

Gene Symbol
TM4SF19-TCTEX1D2
Entrez Gene ID
100534611
Gene Name
TM4SF19-TCTEX1D2 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 3: 196316085 - 196338420 on Build GRCh38
UniGene ID
Hs.733180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037950.1 5 799

View Full Product Details