Gene Transcripts

Gene Symbol
C12orf57
Entrez Gene ID
113246
Gene Name
chromosome 12 open reading frame 57
Gene Aliases
C10, GRCC10
Chromosome Location
Chr. 12: 6943438 - 6946003 on Build GRCh38
UniGene ID
Hs.405913
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301834.1 NP_001288763.1 3, 4 413
NM_001301836.1 NP_001288765.1 2, 3 410
NM_001301838.1 NP_001288767.1 2, 3 638
NM_138425.3 NP_612434.1 2, 3 542
GenBank mRNA AK311912.1 - 2, 3 340
BC009925.2 - 2, 3 319 3537206
BE888740.1 - 2, 3 329
BP318750.2 - 146
CB161655.1 - 3, 4 413
DB494917.1 - 2, 3 410

View Full Product Details