Gene Transcripts

Gene Symbol
GMEB2
Entrez Gene ID
26205
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
GMEB-2, P79PIF, PIF79
Chromosome Location
Chr. 20: 63587594 - 63627101 on Build GRCh38
UniGene ID
Hs.473286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012384.4 NP_036516.1 3, 4 427
XM_005260202.4 XP_005260259.1 3, 4 638
GenBank mRNA AB033095.1 - 3, 4 304
AF173867.1 - 2, 3 703
AK298927.1 - 3, 4 424
BC036305.1 - 3, 4 354 5166938
BQ954281.1 - 2, 3 129
DB474085.1 - 3, 4 380

View Full Product Details