Gene Transcripts

Gene Symbol
NLRP7
Entrez Gene ID
199713
Gene Name
NLR family pyrin domain containing 7
Gene Aliases
CLR19.4, HYDM, NALP7, NOD12, PAN7, PYPAF3
Chromosome Location
Chr. 19: 54923509 - 54965184 on Build GRCh38
UniGene ID
Hs.351118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127255.1 NP_001120727.1 11 3111
NM_139176.3 NP_631915.2 11 3027
NM_206828.3 NP_996611.2 10 2940
XM_006723075.3 XP_006723138.1 12 3346
XM_006723076.3 XP_006723139.1 11 3309
XM_011526596.2 XP_011524898.1 13 3553
XM_011526599.2 XP_011524901.1 13 3564
XM_011526601.2 XP_011524903.1 12 3382
GenBank mRNA AF464765.1 - 9 2910
AK298630.1 - 9 2181
AK302655.1 - 13 3522
AK313964.1 - 13 3506
AY154462.1 - 10 2934
BC109124.2 - 11 3066 40036025
BC109125.1 - 11 3103 40036028

View Full Product Details