Gene Transcripts
- Gene Symbol
- OTOA
- Entrez Gene ID
- 146183
- Gene Name
- otoancorin
- Gene Aliases
- CT108, DFNB22
- Chromosome Location
- Chr. 16: 21663857 - 21771468 on Build GRCh38
- UniGene ID
- Hs.408336
- Species
- Homo sapiens
- Gene Symbol
- LOC653786
- Entrez Gene ID
- 653786
- Gene Name
- otoancorin pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 16: 22545698 - 22576865 on Build GRCh38
- UniGene ID
- Hs.564945
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003676.3 |
|
7 |
2159 |
|
GenBank mRNA |
AK092323.1 |
- |
7 |
2159 |
|
View Full Product Details