Gene Transcripts

Gene Symbol
DHRS13
Entrez Gene ID
147015
Gene Name
dehydrogenase/reductase 13
Gene Aliases
SDR7C5
Chromosome Location
Chr. 17: 28897781 - 28903071 on Build GRCh38
UniGene ID
Hs.631760
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144683.3 NP_653284.2 2, 3 381
GenBank mRNA AY358566.1 - 2, 3 381
BC015582.2 - 1, 2 371 4637504

View Full Product Details