Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144071.1 | NP_001137543.1 | 4 | 576 | |
NM_144972.4 | NP_659409.2 | 3 | 1169 | ||
XM_005252805.4 | XP_005252862.2 | 4 | 458 | ||
XM_011519922.2 | XP_011518224.1 | 4 | 460 | ||
XM_011519923.2 | XP_011518225.1 | 4 | 547 | ||
XM_011519924.2 | XP_011518226.1 | 4 | 404 | ||
XM_011519925.1 | XP_011518227.1 | 4 | 386 | ||
GenBank mRNA | AK131523.1 | - | 3 | 1112 | |
AY581313.1 | - | 4 | 576 | ||
BC014340.1 | - | 4 | 580 | 3935569 |