Gene Transcripts

Gene Symbol
C18orf25
Entrez Gene ID
147339
Gene Name
chromosome 18 open reading frame 25
Gene Aliases
ARKL1, RNF111L1
Chromosome Location
Chr. 18: 46173553 - 46266992 on Build GRCh38
UniGene ID
Hs.116486
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008239.2 NP_001008240.1 2 971
NM_145055.3 NP_659492.1 2 971
XM_011525822.1 XP_011524124.1 2 822
XM_017025555.1 XP_016881044.1 2 818
GenBank mRNA AK289738.1 - 2 858
AK292981.1 - 2 959
AL713661.1 - 1 151
AL832027.2 - 1 499
BC016149.1 - 1 644 4040087
BC084579.1 - 2 938 6141272
BX640683.1 - 1 652

View Full Product Details