Gene Transcripts

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
Chromosome Location
Chr. 5: 181236928 - 181243906 on Build GRCh38
UniGene ID
Hs.5662
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006098.4 NP_006089.1 7 900
GenBank mRNA AK095666.1 - 7 772
AK222488.1 - 7 900
AK301468.1 - 7 826
AY336089.1 - 7 801
BC000214.1 - 7 918 2959178
BC000366.2 - 7 888 2819249
BC010119.2 - 7 875 3835636
BC014256.1 - 7 877 4765908
BC014788.1 - 7 884 3455986
BC017287.2 - 7 865 4892271
BC019093.2 - 7 878 4893346
BC019362.1 - 7 882 2823761
BC021993.1 - 7 900 4083071
BC029996.1 - 7 727
BC032006.1 - 7 879 4750184
BC035460.1 - 7 881
CR456978.1 - 7 794
CR541909.1 - 7 794
GU727632.1 - 7 794
M24194.1 - 7 889

View Full Product Details