Gene Transcripts

Gene Symbol
TMEM237
Entrez Gene ID
65062
Gene Name
transmembrane protein 237
Gene Aliases
ALS2CR4, JBTS14
Chromosome Location
Chr. 2: 201620184 - 201643529 on Build GRCh38
UniGene ID
Hs.12319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044385.2 NP_001037850.1 5 326
NM_152388.3 NP_689601.2 5 491
GenBank mRNA AB053301.1 - 5 493
AK090601.1 - 5 184
AK303954.1 - 5 298
BC029611.1 - 5 239 5304046
BP226834.1 - 5 326
BQ219059.1 - 5 298

View Full Product Details