Gene Transcripts

Gene Symbol
TMCO5A
Entrez Gene ID
145942
Gene Name
transmembrane and coiled-coil domains 5A
Gene Aliases
TMCO5
Chromosome Location
Chr. 15: 37934607 - 37997541 on Build GRCh38
UniGene ID
Hs.179646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152453.3 NP_689666.2 6, 7 585
XM_005254168.3 XP_005254225.1 6, 7 566
XM_011521262.2 XP_011519564.1 6, 7 566
XM_011521263.1 XP_011519565.1 6, 7 566
XM_011521264.1 XP_011519566.1 6, 7 566
GenBank mRNA AK093126.1 - 6, 7 566
AK310517.1 - 6, 7 571
BC029221.2 - 5, 6 498 5167874
BI826726.1 - 6, 7 549

View Full Product Details