Gene Transcripts

Gene Symbol
ZNF48
Entrez Gene ID
197407
Gene Name
zinc finger protein 48
Gene Aliases
ZNF553
Chromosome Location
Chr. 16: 30378312 - 30400108 on Build GRCh38
UniGene ID
Hs.513501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001214906.1 NP_001201835.1 3 278
NM_001214907.1 NP_001201836.1 2 182
NM_001214909.1 NP_001201838.1 3 646
NM_001324494.1 NP_001311423.1 2 552
NM_152652.2 NP_689865.2 2 540
GenBank mRNA AK056313.1 - 2 223
BC041388.1 - 2 227 5276303
BU854776.1 - 3 615
CF594347.1 - 343
DA492655.1 - 2 182
DA508718.1 - 3 280

View Full Product Details