Gene Transcripts

Gene Symbol
VPS37D
Entrez Gene ID
155382
Gene Name
VPS37D, ESCRT-I subunit
Gene Aliases
WBSCR24
Chromosome Location
Chr. 7: 73665346 - 73672110 on Build GRCh38
UniGene ID
Hs.647037
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077621.1 NP_001071089.1 4 1479
XM_017011779.1 XP_016867268.1 4 1669
GenBank mRNA AY081952.1 - 4 1479
BC064621.1 - 4 1326

View Full Product Details