Gene Transcripts

Gene Symbol
BEND7
Entrez Gene ID
222389
Gene Name
BEN domain containing 7
Gene Aliases
C10orf30
Chromosome Location
Chr. 10: 13438484 - 13528772 on Build GRCh38
UniGene ID
Hs.498740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152751.2 NP_689964.2 9 1877
XM_005252402.4 XP_005252459.1 9 1975
XM_011519390.2 XP_011517692.1 8 1668
XM_011519391.2 XP_011517693.1 8 1665
XM_011519392.2 XP_011517694.1 9 1971
XM_011519393.2 XP_011517695.1 8 1763
XM_011519394.2 XP_011517696.1 10 1910
XM_011519395.2 XP_011517697.1 8 2270
XM_011519396.2 XP_011517698.1 9 2361
XM_011519397.2 XP_011517699.1 9 1916
XM_017015915.1 XP_016871404.1 10 1895
GenBank mRNA AK097602.1 - 9 1877

View Full Product Details